missing translation for 'onlineSavingsMsg'
Learn More

(-)-Bicuculline methobromide, Tocris Bioscience™

GABAA antagonist; more water soluble version of (+)-bicuculline (Cat. No. 0130)

Brand:  Tocris (Bio-Techne) 0109/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13467433

  • £114.66 / 10mg

Estimated Shipment date
from Supplier 24-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Methobromide salt of (+)-bicuculline . Water soluble and more stable than bicuculline. Non-GABA receptor-mediated actions reported.

Methoiodide Salt and Methochloride Salt also available.

Specifications

Chemical Identifiers

73604-30-5
462.296
--bicuculline methobromide, 6,6-dimethyl-5-8-oxo-6,8-dihydro-2h-furo 3,4-e 1,3 benzodioxol-6-yl-5,6,7,8-tetrahydro-2h-1,3 dioxolo 4,5-g isoquinolin-6-ium bromide, 6,6-dimethyl-5-12-oxo-3,5,11-trioxatricyclo 7.3.0.0^ 2,6 dodeca-1,6,8-trien-10-yl-2h,5h,7h,8h-1,3 dioxolo 4,5-g isoquinolin-6-ium bromide, 6-6,6-dimethyl-7,8-dihydro-5h-1,3 dioxolo 4,5-g isoquinolin-6-ium-5-yl-6h-furo 3,4-g 1,3 benzodioxol-8-one bromide, 6-6,6-dimethyl-7,8-dihydro-5h-1,3 dioxolo 4,5-g isoquinolin-6-ium-5-yl-6h-furo 3,4-g 1,3 benzodioxol-8-one;bromide
6-(6,6-dimethyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium-5-yl)-6H-furo[3,4-g][1,3]benzodioxol-8-one;bromide
C21H20BrNO6
BWXCECYGGMGBHD-UHFFFAOYSA-M
171729
C[N+]1(CCC2=CC3=C(C=C2C1C4C5=C(C6=C(C=C5)OCO6)C(=O)O4)OCO3)C.[Br-]

Specifications

10mg
73604-30-5
--bicuculline methobromide, 6,6-dimethyl-5-8-oxo-6,8-dihydro-2h-furo 3,4-e 1,3 benzodioxol-6-yl-5,6,7,8-tetrahydro-2h-1,3 dioxolo 4,5-g isoquinolin-6-ium bromide, 6,6-dimethyl-5-12-oxo-3,5,11-trioxatricyclo 7.3.0.0^ 2,6 dodeca-1,6,8-trien-10-yl-2h,5h,7h,8h-1,3 dioxolo 4,5-g isoquinolin-6-ium bromide, 6-6,6-dimethyl-7,8-dihydro-5h-1,3 dioxolo 4,5-g isoquinolin-6-ium-5-yl-6h-furo 3,4-g 1,3 benzodioxol-8-one bromide, 6-6,6-dimethyl-7,8-dihydro-5h-1,3 dioxolo 4,5-g isoquinolin-6-ium-5-yl-6h-furo 3,4-g 1,3 benzodioxol-8-one;bromide
C[N+]1(CCC2=CC3=C(C=C2C1C4C5=C(C6=C(C=C5)OCO6)C(=O)O4)OCO3)C.[Br-]
462.296
462.3
(-)-Bicuculline methobromide
C21H20BrNO6
BWXCECYGGMGBHD-UHFFFAOYSA-M
6-(6,6-dimethyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium-5-yl)-6H-furo[3,4-g][1,3]benzodioxol-8-one;bromide
171729
>98%
Videos
Documents