missing translation for 'onlineSavingsMsg'
Learn More

(-)-Bicuculline methiodide, 98%, Thermo Scientific Chemicals

Catalog Number 15498649
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 40709-69-1
Color Yellow
Molecular Formula C21H20INO6
MDL Number MFCD00078966
Quantity 50 mg
UN Number UN1544
Beilstein 5419649
Synonym --bicuculline methiodide, bicuculline methiodide, 1 s ,9 r---bicuculline methiodide, 5s-5-6r-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo 4,5-g isoquinolinium iodide, 1,3-dioxolo 4,5-g isoquinolinium, 5-6r-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-, iodide, 5s, 1,3-dioxolo 4,5-g isoquinolinium, 5-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-, iodide, r-r*,s*, opera_id_515, --bicucullinemethiodide, 1 s ,9 r---bicuculline methiodide hpce, 5r-5-6s-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo 4,5-g isoquinolinium iodide
InChI Key HKJKCPKPSSVUHY-GRTNUQQKSA-M
SMILES C[N+]1(CCC2=CC3=C(C=C2C1C4C5=C(C6=C(C=C5)OCO6)C(=O)O4)OCO3)C.[I-]
IUPAC Name (6R)-6-[(5S)-6,6-dimethyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium-5-yl]-6H-furo[3,4-g][1,3]benzodioxol-8-one;iodide
Molecular Weight (g/mol) 509.296
PubChem CID 104871
Formula Weight 509.3
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (-)-Bicuculline methiodide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'