missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bicine, 99+%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Absorbance | 0.05 max. (5% aqueous soln.) at 250nm |
Absorbance Conditions | (5% aq. soln.) (1cm cell) |
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Color | White |
pH | 4.0 to 5.5 (10% soln. at 20°C) |
Physical Form | Crystalline Powder |
Quantity | 1 kg |
Assay Percent Range | 99+% |
Infrared Spectrum | Authentic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'