missing translation for 'onlineSavingsMsg'
Learn More

Bicalutamide, Tocris Bioscience™

Non-steroidal androgen receptor antagonist

Brand:  Tocris (Bio-Techne) 338950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13442934

  • £764.40 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Orally active non-steroidal androgen receptor antagonist (IC50 = 190 nM). Displays peripheral selectivity and does not effect serum levels of LH and testosterone. Exhibits potent anticancer activity in vivo.
Specifications

Chemical Identifiers

90357-06-5
430.374
bicalutamide, casodex, cosudex, calutide, bicalutamide cdx, raffolutil, kalumid, casodex tn, bicalutamide usan:inn:ban, bicalutamide casodex
N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide
C18H14F4N2O4S
LKJPYSCBVHEWIU-UHFFFAOYSA-N
2375
CC(CS(=O)(=O)C1=CC=C(C=C1)F)(C(=O)NC2=CC(=C(C=C2)C#N)C(F)(F)F)O

Specifications

50mg
90357-06-5
bicalutamide, casodex, cosudex, calutide, bicalutamide cdx, raffolutil, kalumid, casodex tn, bicalutamide usan:inn:ban, bicalutamide casodex
CC(CS(=O)(=O)C1=CC=C(C=C1)F)(C(=O)NC2=CC(=C(C=C2)C#N)C(F)(F)F)O
430.374
430.37
Bicalutamide
C18H14F4N2O4S
LKJPYSCBVHEWIU-UHFFFAOYSA-N
N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide
2375
>99%
Videos
Documents