missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bialaphos sodium, 85%, Thermo Scientific™
684.90 GBP valid until 2024-03-30
Use promo code "21648" to get your promotional price.
Use promo code "21648" to get your promotional price.
Click to view available options
Quantity:
500mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Bialaphos sodium consists of two L-alanyl units attached to L-phosphinothricyl
• Bialaphos is a bacterial metabolite and an antimicrobial
Application
• Bialaphos is commonly used for transformation experiments in some plant species as a selection agent
• Bialaphos sodium can be used as a standard for metabolite and environmental contaminant determination in laboratory settings
Specifications
UN Number | UN2811 |
Solubility Information | Soluble in water |
Formula Weight | 345.26 |
Percent Purity | 85% |
Color | Orange |
Quantity | 500mg |
Flash Point | >110°C (230°F) |
Boiling Point | 793°C |
Chemical Name or Material | Bialaphos sodium |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'