missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BI 4394

Product Code. 30279875
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30279875 10 mg 10mg
30279870 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30279875 Supplier Tocris Bioscience™ Supplier No. 8842/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent and selective MMP-13 inhibitor

BI 4394 is a highly potent and selective MMP-13 inhibitor (IC50 = 1 nM). BI 4394 inhibits the degradation of isolated bovine nasal cartilage explants (IC50 = 31 nM). It shows high selectivity (>1000 fold) for MMP-13 over other MMPs. In vivo, BI 4394 decreases the mean arthritic score vs control in a murine model of rheumatoid arthritis, when administered at a dose of 100 mg/kg over a 14-day period. BI 4394 is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethyl 5-[3-[[[(4-carboxyphenyl)methyl]amino]carbonyl]-1-methyl-1H-pyrazol-5-yl]-1H-indole-2-carboxylate
CAS 1222173-37-6
Quantity 10 mg
Target Synthetic Metalloprotease Inhibitors
Molecular Formula C24H22N4O5
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.