missing translation for 'onlineSavingsMsg'
Learn More

BGC 20-761, Tocris Bioscience™

High affinity 5-HT6 antagonist

Brand:  Tocris (Bio-Techne) 332650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13442814

  • £487.52 / 50mg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective, high affinity 5-HT6 antagonist (Ki = 20 nM). Reverses the amnesic effects of scopolamine (Cat. No. 1414) and enhances memory consolidation in a rat model.
Specifications

Chemical Identifiers

17375-63-2
294.398
unii-6g5f6esv5i, mpdt, 6g5f6esv5i, chembl7318, 5-methoxy-n,n-dimethyl-2-phenyl-1h-indole-3-ethanamine, 2-5-methoxy-2-phenyl-1h-indol-3-yl-n,n-dimethylethanamine, d0c5gf, gtpl71, 1h-indole-3-ethanamine, 5-methoxy-n,n-dimethyl-2-phenyl, indole, 3-2-dimethylamino ethyl-5-methoxy-2-phenyl
2-(5-methoxy-2-phenyl-1H-indol-3-yl)-N,N-dimethylethanamine
C19H22N2O
VSGPGYWZVPDDSK-UHFFFAOYSA-N
6918515
CN(C)CCC1=C(NC2=C1C=C(C=C2)OC)C3=CC=CC=C3

Specifications

BGC 20-761
17375-63-2
unii-6g5f6esv5i, mpdt, 6g5f6esv5i, chembl7318, 5-methoxy-n,n-dimethyl-2-phenyl-1h-indole-3-ethanamine, 2-5-methoxy-2-phenyl-1h-indol-3-yl-n,n-dimethylethanamine, d0c5gf, gtpl71, 1h-indole-3-ethanamine, 5-methoxy-n,n-dimethyl-2-phenyl, indole, 3-2-dimethylamino ethyl-5-methoxy-2-phenyl
CN(C)CCC1=C(NC2=C1C=C(C=C2)OC)C3=CC=CC=C3
294.398
294.39
50mg
C19H22N2O
VSGPGYWZVPDDSK-UHFFFAOYSA-N
2-(5-methoxy-2-phenyl-1H-indol-3-yl)-N,N-dimethylethanamine
6918515
>98%
Videos