Learn More
BF 2649 hydrochloride, Tocris Bioscience™
H3 receptor inverse agonist/antagonist
Brand:  Tocris (Bio-Techne) 374350
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13433534
  • £627.69 / 50mg
Estimated Shipment date
from Supplier 17-12-2019
Description
Description
Potent and selective histamine H3 receptor inverse agonist (EC50 = 1.5 nM, Ki = 0.16 nM). Exhibits nootropic effects in cognitive disorders; reduces locomotor hyperactivity induced by methamphetamine. Brain penetrant.
Specifications
Chemical Identifiers
903576-44-3
332.309
pitolisant hydrochloride, ciproxidine, unii-yv33ch63hi, yv33ch63hi, 1-3-3-4-chlorophenyl propoxy propyl piperidine hydrochloride, piperidine, 1-3-3-4-chlorophenyl propoxy propyl-, hydrochloride, 1-3-3-4-chlorophenyl propoxy propyl-piperidinehydrochloride
1-[3-[3-(4-chlorophenyl)propoxy]propyl]piperidine;hydrochloride
C17H27Cl2NO
XLFKECRRMPOAQS-UHFFFAOYSA-N
11551689
C1CCN(CC1)CCCOCCCC2=CC=C(C=C2)Cl.Cl
Specifications
50mg
903576-44-3
pitolisant hydrochloride, ciproxidine, unii-yv33ch63hi, yv33ch63hi, 1-3-3-4-chlorophenyl propoxy propyl piperidine hydrochloride, piperidine, 1-3-3-4-chlorophenyl propoxy propyl-, hydrochloride, 1-3-3-4-chlorophenyl propoxy propyl-piperidinehydrochloride
C1CCN(CC1)CCCOCCCC2=CC=C(C=C2)Cl.Cl
332.309
332.31
BF 2649 hydrochloride
C17H27Cl2NO
XLFKECRRMPOAQS-UHFFFAOYSA-N
1-[3-[3-(4-chlorophenyl)propoxy]propyl]piperidine;hydrochloride
11551689
>99%