missing translation for 'onlineSavingsMsg'
Learn More

Betulinic acid, Tocris Bioscience™

Antitumor and anti-HIV agent; activates NF-κB. Also TGR5 agonist

Brand:  Tocris (Bio-Techne) 390650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13443794

  • £286.65 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Natural triterpenoid that displays anti-HIV and antitumor activity. Induces the production of reactive oxygen species (ROS) and activates NF-κB. Also a GPBA receptor partial agonist (EC50 = 1.04 μM, efficacy 83%).
Specifications

Chemical Identifiers

472-15-1
456.711
betulinic acid, mairin, betulic acid, ccris 6748, 3-hydroxylup-20 29-en-28-oic acid, 3beta-hydroxy-20 29-lupaene-28-oic acid, 3beta-hydroxy-lup-20 29-en-28-oic acid, lup-20 29-en-28-oic acid, 3-hydroxy-, 3beta, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-1-isopropenyl-5a,5b,8,8,11a-pentamethyl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid
CHEBI:3087
CC(=C)C1CCC2(C1C3CCC4C5(CCC(C(C5CCC4(C3(CC2)C)C)(C)C)O)C)C(=O)O
C30H48O3
QGJZLNKBHJESQX-FZFNOLFKSA-N
64971
(1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysene-3a-carboxylic acid

Specifications

50mg
472-15-1
betulinic acid, mairin, betulic acid, ccris 6748, 3-hydroxylup-20 29-en-28-oic acid, 3beta-hydroxy-20 29-lupaene-28-oic acid, 3beta-hydroxy-lup-20 29-en-28-oic acid, lup-20 29-en-28-oic acid, 3-hydroxy-, 3beta, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-1-isopropenyl-5a,5b,8,8,11a-pentamethyl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid
CC(=C)C1CCC2(C1C3CCC4C5(CCC(C(C5CCC4(C3(CC2)C)C)(C)C)O)C)C(=O)O
456.711
CHEBI:3087
>98%
Betulinic acid
C30H48O3
QGJZLNKBHJESQX-FZFNOLFKSA-N
(1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysene-3a-carboxylic acid
64971
456.7
Videos
Documents