missing translation for 'onlineSavingsMsg'
Learn More

beta-Propiolactone, 97%, Thermo Scientific Chemicals

Catalog Number p-7026898
Click to view available options
Quantity:
1 g
5 g
25 g
57-57-8
C3H4O2
72.06
MFCD00005169
VEZXCJBBBCKRPI-UHFFFAOYSA-N
beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolakton
2365
CHEBI:49073
oxetan-2-one
O=C1CCO1
57-57-8
C3H4O2
72.06
MFCD00005169
VEZXCJBBBCKRPI-UHFFFAOYSA-N
beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolakton
2365
CHEBI:49073
oxetan-2-one
O=C1CCO1

Beta-Propiolactone is widely used as a sterilizing and sporicidal agent, which is used to sterilize blood plasma, vaccines, tissue grafts, surgical instruments and enzymes. It finds an application in the manufacturing of acrylic acid and its esters. It is utilized as an intermediate in the synthesis of synthetic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Beta-Propiolactone is widely used as a sterilizing and sporicidal agent, which is used to sterilize blood plasma, vaccines, tissue grafts, surgical instruments and enzymes. It finds an application in the manufacturing of acrylic acid and its esters. It is utilized as an intermediate in the synthesis of synthetic compounds.

Solubility
Miscible with water and organic solvents.

Notes
Flammable material. Should be stored in a separate safety storage cabinet or room. Keep away from heat and sources of ignition.
TRUSTED_SUSTAINABILITY
Melting Point -34°C
Density 1.146
Boiling Point 161°C to 163°C
Flash Point 70°C (158°F)
Assay Percent Range 0.97
Refractive Index 1.413
Quantity 1 g
UN Number UN3382
Sensitivity Moisture sensitive
Merck Index 14,7820
Solubility Information Miscible with water and organic solvents.
Formula Weight 72.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material beta-Propiolactone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'