missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide adenine dinucleotide, disodium salt, hydrate, 95+%, reduced form, Thermo Scientific Chemicals

Catalog Number 10711911
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
1949720-50-6
C21H27N7Na2O14P2
709.41
MFCD00036200
QRGNQKGQENGQSE-WUEGHLCSSA-L
beta-nadh disodium salt
131673989
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium
[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
1949720-50-6
C21H27N7Na2O14P2
709.41
MFCD00036200
QRGNQKGQENGQSE-WUEGHLCSSA-L
beta-nadh disodium salt
131673989
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium
[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance 0.50mg/mL NADH in 10mM TRIS solution
CAS Min % 95.0
CAS Max % 100.0
Melting Point 140.0°C to 142.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 709.4
Percent Purity 95+%
Physical Form Powder
Chemical Name or Material β-Nicotinamide adenine dinucleotide, disodium salt, hydrate, Reduced Form
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'