missing translation for 'onlineSavingsMsg'
Learn More

β-Glycerophosphoric acid, disodium salt pentahydrate, 98%, Thermo Scientific Chemicals

Catalog Number 10561814
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13408-09-8
C3H19Na2O11P
308.13
MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135
VWVLGHQKEJESPW-UHFFFAOYSA-N
unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate
131846916
1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate
O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O
This item is not returnable. View return policy
13408-09-8
C3H19Na2O11P
308.13
MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135
VWVLGHQKEJESPW-UHFFFAOYSA-N
unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate
131846916
1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate
O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O

beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%, C3H7Na2O6P.5H2O, CAS Number-13408-09-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 102.0°C to 104.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH2OHCH(OPO3Na2)CH2OH·5H2O
Quantity 500 g
Solubility Information Solubility in water: freely soluble.
Formula Weight 306.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material β-Glycerophosphoric acid, disodium salt pentahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'