missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-D-Glucosamine pentaacetate, 96%

Catalog Number p-7044993
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7772-79-4
C16H23NO10
389.357
MFCD00006595
OVPIZHVSWNOZMN-OXGONZEZSA-N
n-acetyl-beta-d-glucosamine tetraacetate, 2-acetamido-1,3,4,6-tetra-o-acetyl-2-deoxy-beta-d-glucopyranose, beta-d-glucosamine pentaacetate, 2-acetamido-2-deoxy-beta-d-glucopyranose 1,3,4,6-tetraacetate, 2r,3s,4r,5r,6s-5-acetamido-3,4,6-triacetoxy-tetrahydropyran-2-yl methyl acetate, beta-d-galactosamine pentaacetate, 2-acetylamino-2-deoxy-beta-d-galactopyranose 1,3,4,6-tetraacetate, 2r,3s,4r,5r,6s-3,4,6-tris acetyloxy-5-acetamidooxan-2-yl methyl acetate, n-acetyl-, a-d-glucosamine tetraacetate, glc-n-ac free
99461
[(2R,3S,4R,5R,6S)-5-acetamido-3,4,6-triacetyloxyoxan-2-yl]methyl acetate
CC(=O)NC1C(C(C(OC1OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C
This item is not returnable. View return policy
7772-79-4
C16H23NO10
389.357
MFCD00006595
OVPIZHVSWNOZMN-OXGONZEZSA-N
n-acetyl-beta-d-glucosamine tetraacetate, 2-acetamido-1,3,4,6-tetra-o-acetyl-2-deoxy-beta-d-glucopyranose, beta-d-glucosamine pentaacetate, 2-acetamido-2-deoxy-beta-d-glucopyranose 1,3,4,6-tetraacetate, 2r,3s,4r,5r,6s-5-acetamido-3,4,6-triacetoxy-tetrahydropyran-2-yl methyl acetate, beta-d-galactosamine pentaacetate, 2-acetylamino-2-deoxy-beta-d-galactopyranose 1,3,4,6-tetraacetate, 2r,3s,4r,5r,6s-3,4,6-tris acetyloxy-5-acetamidooxan-2-yl methyl acetate, n-acetyl-, a-d-glucosamine tetraacetate, glc-n-ac free
99461
[(2R,3S,4R,5R,6S)-5-acetamido-3,4,6-triacetyloxyoxan-2-yl]methyl acetate
CC(=O)NC1C(C(C(OC1OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 184°C to 189°C
Quantity 1 g
Beilstein 98835
Optical Rotation +4° (c=10 in Chloroform)
Formula Weight 389.37
Percent Purity 96%
Chemical Name or Material beta-D-Glucosamine pentaacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'