missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-D-Galactose pentaacetate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It is used as a pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediates.
Solubility
Soluble in methanol (50 mg/ml), chloroform, and water.
Notes
Store at room temperature. Store away from oxidizing agent.
It is used as a pharmaceutical intermediates.
Solubility
Soluble in methanol (50 mg/ml), chloroform, and water.
Notes
Store at room temperature. Store away from oxidizing agent.
Specifications
Melting Point | 142°C to 146°C |
Quantity | 5 g |
Beilstein | 98853 |
Solubility Information | Soluble in methanol (50mg/ml),chloroform,and water. |
Optical Rotation | +24° (c=4 in chloroform) |
Formula Weight | 390.34 |
Percent Purity | 98% |
Chemical Name or Material | beta-D-Galactose pentaacetate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'