missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta Catenin Mouse, Clone: CTNNB1/1509, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | Beta Catenin |
---|---|
Clone | CTNNB1/1509 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939877
|
EnquireBio
1499-MSM9-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929877
|
EnquireBio
1499-MSM9-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Beta Catenin | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
1499 | |
Recombinant human full-length beta-Catenin (p120) protein fragment | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
92kDa |
CTNNB1/1509 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001895 | |
CTNNB1 | |
IgG1 κ | |
Protein A/G Purified | |
Cell surface and cytoplasmic | |
HeLa or MCF-7 cells. Breast carcinoma |