missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Carotene, 99%, Thermo Scientific Chemicals
18.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
Description
The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro.
Solubility
Soluble in hexane, dimethyl sulfoxide, benzene, chloroform, cyclohexane. Insoluble in water.
Notes
Air & Light Sensitive. Store in a cool place.
The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro.
Solubility
Soluble in hexane, dimethyl sulfoxide, benzene, chloroform, cyclohexane. Insoluble in water.
Notes
Air & Light Sensitive. Store in a cool place.
Specifications
Melting Point | 178°C to 179°C |
Density | 1.000 g/mL |
Flash Point | 103°C (217°F) |
Quantity | 1 g |
Beilstein | 1917416 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in hexane,dimethyl sulfoxide,benzene,chloroform,cyclohexane. Insoluble in water. |
Formula Weight | 536.87 |
Percent Purity | 99% |
Physical Form | Solid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'