missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Alanine tert-butyl ester hydrochloride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Useful intermediate for pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Useful intermediate for pharmaceutical and organic synthesis.
Solubility
Clear solution (0.3g in 2ml MeOH).
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Useful intermediate for pharmaceutical and organic synthesis.
Solubility
Clear solution (0.3g in 2ml MeOH).
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
Melting Point | 167°C to 169°C |
Linear Formula | H2NCH2CH2COOC(CH3)3·HCl |
Beilstein | 6660122 |
Solubility Information | Clear solution (0.3g in 2ml MeOH). |
Formula Weight | 181.66 |
Percent Purity | 95% |
Quantity | 25 g |
Physical Form | Solid |
Chemical Name or Material | beta-Alanine tert-butyl ester hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'