missing translation for 'onlineSavingsMsg'
Learn More

β-Alanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number 10591892
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
3196-73-4
C4H10ClNO2
139.58
MFCD00039060
XPGRZDJXVKFLHQ-UHFFFAOYSA-N
methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride
2734767
methyl 3-aminopropanoate;hydrochloride
Cl.COC(=O)CCN
This item is not returnable. View return policy
3196-73-4
C4H10ClNO2
139.58
MFCD00039060
XPGRZDJXVKFLHQ-UHFFFAOYSA-N
methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride
2734767
methyl 3-aminopropanoate;hydrochloride
Cl.COC(=O)CCN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 103.0°C to 105.0°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Assay Percent Range 98%
Infrared Spectrum Authentic
Linear Formula NH2CH2CH2COOCH3·HCl
Beilstein 04, IV, 2527
Packaging Glass Bottle
Formula Weight 139.58
Percent Purity 98%
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material β-Alanine methyl ester hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'