missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals β-Alanine, 99%

Catalog Number p-3776557
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy
107-95-9
C3H7NO2
89.09
MFCD00008200
UCMIRNVEIXFBKS-UHFFFAOYSA-N
beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
239
CHEBI:16958
3-aminopropanoic acid
NCCC(O)=O
This item is not returnable. View return policy
107-95-9
C3H7NO2
89.09
MFCD00008200
UCMIRNVEIXFBKS-UHFFFAOYSA-N
beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
239
CHEBI:16958
3-aminopropanoic acid
NCCC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 197.0°C to 202.0°C
Ammonium 1000ppm max.
CAS Min % 98.5
CAS Max % 100.0
pH 6.5 to 7.5 (10% w/v aq. soln.)
Color White
Chlorine (Cl) 400ppm max.
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 99%
Infrared Spectrum Authentic
Linear Formula H2NCH2CH2CO2H
Iron (Fe) 30ppm max.
Fieser 01,16; 05,6; 06,10; 17,3
Loss on Drying 0.5% max. (105°C, 3 hrs)
Other Amino Acids (TLC) Passes Test (not detectable on 10μgspotting)
Packaging Plastic Bottle
Sulfate 480ppm max.
Formula Weight 89.09
Percent Purity 99%
Sulfated Ash 0.2% max.
Quantity 500 g
Physical Form Crystalline Powder
Chemical Name or Material β-Alanine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'