Learn More
SHOWA™ N-DEX™ Original and N-DEX Plus Nitrile Gloves
Three times more resistant to punctures than latex and vinyl
Brand:  SHOWA™ BST8005PF2
UNSPSC : 46181504
Code : S3
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13147554
  • £46.75 / Pack of 50
In Stock 6
Add to basket
Specifications Description & Specifications
Specifications
Smooth
Rolled
Yes
Medium
Powder-free
24cm
Yes
Ambidextrous
Nitrile
21 CFR 170-199, FDA, NFPA, OSHA
Blue
Documentation