Learn More
Thermo Scientific Alfa Aesar Beryllium wire, 1.5mm (0.060 in.) dia., 98.5% (metals basis)
Brand: Thermo Scientific Alfa Aesar 044795.BQ
Beryllium wire, 1.5mm (0.060 in.) dia., 98.5% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-41-7 Weight : 0.01000kg