Learn More
Alfa Aesar™ Beryllium wire, 1.5mm (0.060 in.) dia., 98.5% (metals basis)
Brand: Alfa Aesar™ 044795.BS
Code : NEW
Additional Details : CAS Number : 7440-41-7 Weight : 0.01000kg
Quantity | 10cm |
---|
Chemical Identifiers
Beryllium wire | |
7440-41-7 | |
1.5mm (0.060 in.) dia., Temper: as drawn | |
MFCD00134032 |
Specifications
Beryllium wire | |
10cm | |
7440-41-7 | |
98.5% | |
MFCD00134032 |
≈3.3g/m | |
1.5mm (0.060 in.) dia., Temper: as drawn | |
Wire | |
(metals basis) | |
Odorless |