Learn More
Thermo Scientific Alfa Aesar Beryllium wire, 0.05mm (0.002 in.) dia., annealed, 99.7% (metals basis)
Brand: Thermo Scientific Alfa Aesar 040681.BU
Beryllium wire, 0.05mm (0.002 in.) dia., annealed, 99.7% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-41-7 Weight : 0.01000kg