missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Beryllium wire, 0.05mm (0.002 in.) dia., annealed, 99.7% (metals basis)

Alloy source, X-ray tube manufacture, neutron source

Brand:  Alfa Aesar™ 040681.G1

Code : NEW

Additional Details : CAS Number : 7440-41-7 Weight : 0.01000kg

Quantity 1m

 View more versions of this product

Product Code. 11378618

  • £309.63 / 1m


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

Beryllium wire
7440-41-7
0.05mm (0.002 in.) dia., Annealed, Temper: soft
MFCD00134032

Specifications

Beryllium wire
1m
7440-41-7
99.7%
MFCD00134032
≈0.004g/m
0.05mm (0.002 in.) dia., Annealed, Temper: soft
Wire
(metals basis)
Odorless
Videos
SDS