Learn More
Thermo Scientific Alfa Aesar Beryllium Thinfoil, 0.01mm (0.0004 in.) thick, 99.0% (metals basis)
Brand: Thermo Scientific Alfa Aesar 041611.FF
Beryllium Thinfoil, 0.01mm (0.0004 in.) thick, 99.0% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-41-7 Weight : 0.01000kg