missing translation for 'onlineSavingsMsg'
Learn More

Beryllium, plasma standard solution, Specpure™ Be 10,000μg/mL, Thermo Scientific Chemicals

Catalog Number 11312338
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
MFCD00134032
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
MFCD00134032

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration Be4O(C2H3O2)6 in 5% HNO3
Quantity 100 mL
UN Number UN2922
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Beryllium plasma standard solution, Be 10000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'