Learn More
Thermo Scientific Alfa Aesar Beryllium foil, 0.05mm (0.002 in.) thick, 99.8% (metals basis)
Brand: Thermo Scientific Alfa Aesar 041614.FF
Beryllium foil, 0.05mm (0.002 in.) thick, 99.8% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-41-7 Weight : 0.01000kg