Learn More
Berkshire™ SatPax™ 1000 Presaturated Wipers
For ISO Class 5 to 8 (100 to 100000) Cleanroom Class applications
Brand: Berkshire™ SPX1000.001.12
Code : 04
Additional Details : CAS Number : 67-63-0 Weight : 0.80000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
- Nonwoven hydroentangled 55% cellulose/45% polyester construction
- Presaturated to eliminate the need to store, mix, and dispense solvents—no more squirt bottles or uncontrolled saturation levels
- Manufactured using isopropyl alcohol and reverse osmosis/deionized water is filtered to 0.2μm — two saturation concentrations available
Order Info
Packaged in resealable pouches with lot numbers for traceability and canister dispensers with perforated rolls for popup convenience; canister refills available. Basis weight: 67.6g/cm2.
Specifications
SatPax™ 1000 Pre-saturated Resealable Pouch | |
22.86 cm | |
22.86 cm | |
67.6 | |
Knife-cut | |
For ISO Class 5 to 8 applications, presaturated to eliminate the need to store, mix, and dispense solvents no more squirt bottles or uncontrolled saturation levels | |
70% Isopropyl Alcohol, 30% Deionised Water |
9 in. | |
9 in. | |
Polyester Cellulose | |
White | |
9 x 9 in. (22.86 x 22.86 cm) | |
75/Pk. | |
Non-sterile |
Safety and Handling
- Satpax 70% IPA/30% DI H2O
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol