missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Berkshire™ BCR™ Polyester Half-Finger Cleanroom Glove Liners
Continuous filament knitted polyester construction is thin yet durable, while the half-finger design permits tactility in addition to wicking moisture from the palms.
Brand: Berkshire™ BGL2.20L
Code : 04
Additional Details : Weight : 0.08640kg
Description
- Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
- Half-finger design permits tactility while also wicking moisture from the palms
- 100% continuous filament knitted polyester is thin, yet durable
- Ambidextrous design can be used on either hand
- Gloves can be repeatedly laundered for reuse
- Suitable for use in ISO Class Cleanrooms
Specifications
Uncoated | |
Reusable | |
Polyester | |
Half | |
Knit | |
Pack |
White | |
Ambidextrous | |
Large | |
Laboratories and Manufacturing Environments | |
Blue | |
BGL2 |