missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriphenylphosphonium Chloride, 99%, Thermo Scientific Chemicals

Catalog Number 10696394
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1100-88-5
C25H22ClP
388.87
MFCD00011913
USFRYJRPHFMVBZ-UHFFFAOYSA-M
benzyltriphenylphosphonium chloride, triphenylbenzylphosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride, benzyltriphenylphosphanium chloride, benzyl triphenylphosphonium chloride, benzyl-triphenyl-phosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride 1:1, triphenylbenzylphosphine, chloride, benzyl triphenyl phosphoniumchlorid
70671
benzyl(triphenyl)phosphanium;chloride
[Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1100-88-5
C25H22ClP
388.87
MFCD00011913
USFRYJRPHFMVBZ-UHFFFAOYSA-M
benzyltriphenylphosphonium chloride, triphenylbenzylphosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride, benzyltriphenylphosphanium chloride, benzyl triphenylphosphonium chloride, benzyl-triphenyl-phosphonium chloride, phosphonium, triphenyl phenylmethyl-, chloride 1:1, triphenylbenzylphosphine, chloride, benzyl triphenyl phosphoniumchlorid
70671
benzyl(triphenyl)phosphanium;chloride
[Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 337.0°C
Color White
Flash Point >300°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H5CH2P(C6H5)3Cl
Quantity 500 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,slightly soluble in lower aliphatic alcohols
Formula Weight 388.86
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Benzyltriphenylphosphonium chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzyltriphenylphosphonium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'