missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltriphenylphosphonium borohydride, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Reduction reagent
Specifications
Melting Point | 157.0°C to 163.0°C |
Assay Percent Range | 95% |
Packaging | Glass bottle |
Quantity | 5 g |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in chloroform,dichloromethane,insoluble in benzene and hexane |
Formula Weight | 368.27 |
Percent Purity | 95% |
Chemical Name or Material | Benzyltriphenylphosphonium borohydride |
Safety and Handling
Product Identifier
- Benzyltriphenylphosphonium borohydride
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 2
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H261-In contact with water releases flammable gases.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'