missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Benzyltrimethylammonium hydroxide, 40% w/w in methanol

£13.30 - £169.36

Chemical Identifiers

CAS 100-85-6
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00008281
InChI Key NDKBVBUGCNGSJJ-UHFFFAOYSA-M
Synonym benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
PubChem CID 66854
IUPAC Name benzyl(trimethyl)azanium;hydroxide
SMILES C[N+](C)(C)CC1=CC=CC=C1.[OH-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11442747 SDS View Documents Alfa Aesar™
A14927.AE
100mL £13.30
100mL View alternative products
Save Up to 
»
EU Stock 11 Please to purchase this item. Need a web account? Register with us today!
11452747 SDS View Documents Alfa Aesar™
A14927.AP
500mL £43.95
500mL View alternative products
Save Up to 
»
EU Stock 27 Please to purchase this item. Need a web account? Register with us today!
11462747 SDS View Documents Alfa Aesar™
A14927.0F
2500mL £169.36
2500mL View alternative products
Save Up to 
»
EU Stock 14 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications
Benzyltrimethylammonium hydroxide is used as a phase-transfer catalyst. It is also used in aldol condensation reactions and base-catalyzed dehydration reactions. Further, it serves as a strong organic base and used in Horner-Wadsworth-Emmons olefination reactions. In addition, it is an active component in the formulation, which is used in silicon wafer cleaning applications.

Solubility
Miscible with water, hydrocarbons, aromatic hydrocarbons and halogenated solvents.

Notes
Air Sensitive. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

100-85-6
167.252
NDKBVBUGCNGSJJ-UHFFFAOYSA-M
66854
C[N+](C)(C)CC1=CC=CC=C1.[OH-]
C10H17NO
MFCD00008281
benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
benzyl(trimethyl)azanium;hydroxide

Specifications

Benzyltrimethylammonium hydroxide
15°C (59°F)
1.427
65°C
100-85-6
C6H5CH2N(CH3)3OH
UN2924
benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
Miscible with water,hydrocarbons,aromatic hydrocarbons and halogenated solvents.
C[N+](C)(C)CC1=CC=CC=C1.[OH-]
167.252
167.25
0.92
Ammonia-like
Air Sensitive
100mL
C10H17NO
MFCD00008281
3917256
40% w/w in methanol
NDKBVBUGCNGSJJ-UHFFFAOYSA-M
benzyl(trimethyl)azanium;hydroxide
66854