missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltrimethylammonium hydroxide, 40% w/w in methanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
Benzyltrimethylammonium hydroxide is used as a phase-transfer catalyst. It is also used in aldol condensation reactions and base-catalyzed dehydration reactions. Further, it serves as a strong organic base and used in Horner-Wadsworth-Emmons olefination reactions. In addition, it is an active component in the formulation, which is used in silicon wafer cleaning applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzyltrimethylammonium hydroxide is used as a phase-transfer catalyst. It is also used in aldol condensation reactions and base-catalyzed dehydration reactions. Further, it serves as a strong organic base and used in Horner-Wadsworth-Emmons olefination reactions. In addition, it is an active component in the formulation, which is used in silicon wafer cleaning applications.
Solubility
Miscible with water, hydrocarbons, aromatic hydrocarbons and halogenated solvents.
Notes
Air Sensitive. Incompatible with strong oxidizing agents.
Benzyltrimethylammonium hydroxide is used as a phase-transfer catalyst. It is also used in aldol condensation reactions and base-catalyzed dehydration reactions. Further, it serves as a strong organic base and used in Horner-Wadsworth-Emmons olefination reactions. In addition, it is an active component in the formulation, which is used in silicon wafer cleaning applications.
Solubility
Miscible with water, hydrocarbons, aromatic hydrocarbons and halogenated solvents.
Notes
Air Sensitive. Incompatible with strong oxidizing agents.
Specifications
Density | 0.92 |
Boiling Point | 65°C |
Flash Point | 15°C (59°F) |
Odor | Ammonia-like |
Linear Formula | C6H5CH2N(CH3)3OH |
Refractive Index | 1.427 |
Quantity | 100 mL |
UN Number | UN2924 |
Beilstein | 3917256 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'