missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium hydroxide, 20% w/w aq. soln., Thermo Scientific™

Catalog Number p-7051652
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy
100-85-6
C10H17NO
167.252
MFCD00008281
NDKBVBUGCNGSJJ-UHFFFAOYSA-M
benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
66854
benzyl(trimethyl)azanium;hydroxide
C[N+](C)(C)CC1=CC=CC=C1.[OH-]
This item is not returnable. View return policy
100-85-6
C10H17NO
167.252
MFCD00008281
NDKBVBUGCNGSJJ-UHFFFAOYSA-M
benzyltrimethylammonium hydroxide, triton b, n,n,n-trimethyl-1-phenylmethanaminium hydroxide, trimethylbenzylammonium hydroxide, benzyl trimethylammonium hydroxide, benzyl trimethyl ammonium hydroxide, trimethyl benzylammonium hydroxide, sumquat 2311, benzyltrimetylammonium hydroxide, n,n,n-trimethylbenzenemethanaminium hydroxide
66854
benzyl(trimethyl)azanium;hydroxide
C[N+](C)(C)CC1=CC=CC=C1.[OH-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Odor Amine-like
Linear Formula C6H5CH2N(CH3)3OH
Quantity 250 mL
UN Number UN3267
Sensitivity Air Sensitive
Formula Weight 167.25
Concentration or Composition (by Analyte or Components) 20% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Benzyltrimethylammonium hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'