missing translation for 'onlineSavingsMsg'
Learn More

Benzyltriethylammonium bromide, 99%, Thermo Scientific Chemicals

Catalog Number p-66103
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5197-95-5
C13H22BrN
272.23
MFCD00011822
CHQVQXZFZHACQQ-UHFFFAOYSA-M
benzyltriethylammonium bromide, n-benzyl-n,n-diethylethanaminium bromide, benzyl triethyl ammonium bromide, triethylbenzylammonium bromide, bteab, benzenemethanaminium, n,n,n-triethyl-, bromide, benzyltriethyl ammonium bromide, n-benzyl-n,n,n-triethylammoniumbromide, benzyltriethylazanium bromide, pubchem7797
165294
benzyl(triethyl)azanium;bromide
[Br-].CC[N+](CC)(CC)CC1=CC=CC=C1
This item is not returnable. View return policy
5197-95-5
C13H22BrN
272.23
MFCD00011822
CHQVQXZFZHACQQ-UHFFFAOYSA-M
benzyltriethylammonium bromide, n-benzyl-n,n-diethylethanaminium bromide, benzyl triethyl ammonium bromide, triethylbenzylammonium bromide, bteab, benzenemethanaminium, n,n,n-triethyl-, bromide, benzyltriethyl ammonium bromide, n-benzyl-n,n,n-triethylammoniumbromide, benzyltriethylazanium bromide, pubchem7797
165294
benzyl(triethyl)azanium;bromide
[Br-].CC[N+](CC)(CC)CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 187.0°C to 197.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H5CH2N(C2H5)3Br
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 272.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Benzyltriethylammonium bromide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Benzyltriethylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'