missing translation for 'onlineSavingsMsg'
Learn More

Benzyltributylammonium bromide, 99%, Thermo Scientific™

Catalog Number p-66068
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
25316-59-0
MFCD00011848
UDYGXWPMSJPFDG-UHFFFAOYSA-M
benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801
2724282
benzyl(tributyl)azanium;bromide
CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]
This item is not returnable. View return policy
25316-59-0
MFCD00011848
UDYGXWPMSJPFDG-UHFFFAOYSA-M
benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801
2724282
benzyl(tributyl)azanium;bromide
CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 168.0°C to 174.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C19H34BrN
Linear Formula C6H5CH2N[(CH2)3CH3]3Br
Quantity 5 g
Molecular Weight (g/mol) 356.38
Formula Weight 356.38
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Benzyltributylammonium bromide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Benzyltributylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'