missing translation for 'onlineSavingsMsg'
Learn More

Benzyltri-n-butylammonium chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7080704
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
23616-79-7
C19H34ClN
311.94
MFCD00011849
VJGNLOIQCWLBJR-UHFFFAOYSA-M
benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac
159952
benzyl(tributyl)azanium;chloride
[Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1
This item is not returnable. View return policy
23616-79-7
C19H34ClN
311.94
MFCD00011849
VJGNLOIQCWLBJR-UHFFFAOYSA-M
benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac
159952
benzyl(tributyl)azanium;chloride
[Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 156°C to 162°C
Quantity 100 g
Beilstein 3776210
Formula Weight 311.94
Percent Purity 98%
Chemical Name or Material Benzyltri-n-butylammonium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'