missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetaldehyde, 95%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3598310
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
60656-87-3
C9H10O2
150.18
MFCD00191779
NFNOAHXEQXMCGT-UHFFFAOYSA-N
2-benzyloxy acetaldehyde, benzyloxyacetaldehyde, benzyloxy acetaldehyde, phenylmethoxy acetaldehyde, acetaldehyde, phenylmethoxy, acetaldehyde,2-phenylmethoxy, acmc-20ahpl, benzyloxy-acetaldehyde, 2-phenylmethoxyethanal
108989
O=CCOCC1=CC=CC=C1
This item is not returnable. View return policy
60656-87-3
C9H10O2
150.18
MFCD00191779
NFNOAHXEQXMCGT-UHFFFAOYSA-N
2-benzyloxy acetaldehyde, benzyloxyacetaldehyde, benzyloxy acetaldehyde, phenylmethoxy acetaldehyde, acetaldehyde, phenylmethoxy, acetaldehyde,2-phenylmethoxy, acmc-20ahpl, benzyloxy-acetaldehyde, 2-phenylmethoxyethanal
108989
O=CCOCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.0690g/mL
Boiling Point 118°C to 120°C (13.0 mmHg)
Flash Point 113°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula C6H5CH2OCH2CHO
Quantity 1 g
Beilstein 06,III,1476
Specific Gravity 1.069
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chloroform
IUPAC Name 2-phenylmethoxyacetaldehyde
Formula Weight 150.18
Percent Purity 95%
Chemical Name or Material Benzyloxyacetaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'