missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylmagnesium chloride, 1.5M solution in THF, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown to Black |
CAS Min % | 20.51, 75.83 |
CAS Max % | 24.17, 79.49 |
Linear Formula | (C6H5)CH2MgCl |
Concentration | 1.40 to 1.65M |
Density | 1.0300g/mL |
Solubility Information | Solubility in water: reacts. |
Formula Weight | 150.89 |
Percent Purity | 19 to 21% (as BMC) (Total base) |
Packaging | AcroSeal™ Glass Bottle |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'