missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethylhexadecylammonium chloride hydrate, 95%, Thermo Scientific Chemicals

Catalog Number p-7049971
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
122-18-9
C25H46ClN
396.10
MFCD00149967
SXPWTBGAZSPLHA-UHFFFAOYSA-M
cetalkonium chloride, benzyldimethylhexadecylammonium chloride, banicol, bonjela, cetyl zephiran, cetyldimethylbenzylammonium chloride, bicetonium, acinol, benzylhexadecyldimethylammonium chloride, baktonium
31202
benzyl-hexadecyl-dimethylazanium;chloride
[Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
This item is not returnable. View return policy
122-18-9
C25H46ClN
396.10
MFCD00149967
SXPWTBGAZSPLHA-UHFFFAOYSA-M
cetalkonium chloride, benzyldimethylhexadecylammonium chloride, banicol, bonjela, cetyl zephiran, cetyldimethylbenzylammonium chloride, bicetonium, acinol, benzylhexadecyldimethylammonium chloride, baktonium
31202
benzyl-hexadecyl-dimethylazanium;chloride
[Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Benzyldimethylhexadecylammonium chloride has been used in a study to assess the removal of cationic surfactants from aqueous solutions using activated carbon cloth (ACC). It has also been used in a study to investigate the prototropic behavior of the dye, 7-hydroxy-4-methylcoumarin (7H4MC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 54°C to 58°C
Color White
Linear Formula CH3(CH2)15N(CH2C6H5)(CH3)2Cl·xH2O
Quantity 10 g
UN Number UN3261
Beilstein 3802341
Merck Index 14,2017
Formula Weight 396.10 (Anhydrous)
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Benzyldimethylhexadecylammonium chloride hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'