missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate, 97%, ACROS Organics™

Brand:  Acros Organics 434190050

Code : 29

Additional Details : CAS Number : 87219-29-2 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 10301730

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate
96.0
97%
C12H13NO4
MFCD00216569
87219-29-2
100.0
97%
235.24

Specifications

Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate
96.0
Authentic
Crystalline Powder
97%
C12H13NO4
105.0° to 106.5°C
White
87219-29-2
100.0
Glass bottle
−47° to −57° (20°C, 589nm) (C=1, acetone)
97%
235.24
MFCD00216569
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate

Signal Word
  • Warning

Hazard Category
  • Respiratory sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

SDS