missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate, 97%, ACROS Organics™

Brand:  Acros Organics 434190010

Code : 29

Additional Details : CAS Number : 87219-29-2 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 10129699

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate
96.0
≥96.0% (HPLC)
C12H13NO4
MFCD00216569
87219-29-2
100.0
97%
235.24

Specifications

Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate
96.0
Authentic
−47° to −57° (20°C, 589nm) (C=1, acetone)
97%
235.24
MFCD00216569
1g
87219-29-2
100.0
Crystalline Powder
≥96.0% (HPLC)
C12H13NO4
103° to 108°C
White
Videos
SDS