missing translation for 'onlineSavingsMsg'
Learn More

Benzyl phenyl ether, 98%, Thermo Scientific Chemicals

Catalog Number 10635531
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
946-80-5
C13H12O
184.24
MFCD00020660
BOTNYLSAWDQNEX-UHFFFAOYSA-N
benzyl phenyl ether, benzyloxy benzene, phenyl benzyl ether, benzene, phenoxymethyl, ether, benzyl phenyl, benzylphenylether, benzyloxy-benzene, benzyloxybenzene, unii-bue863n0l8, .alpha.-phenylanisole
70352
phenoxymethylbenzene
C1=CC=C(C=C1)COC2=CC=CC=C2
This item is not returnable. View return policy
946-80-5
C13H12O
184.24
MFCD00020660
BOTNYLSAWDQNEX-UHFFFAOYSA-N
benzyl phenyl ether, benzyloxy benzene, phenyl benzyl ether, benzene, phenoxymethyl, ether, benzyl phenyl, benzylphenylether, benzyloxy-benzene, benzyloxybenzene, unii-bue863n0l8, .alpha.-phenylanisole
70352
phenoxymethylbenzene
C1=CC=C(C=C1)COC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 41°C
Color Brown-Orange
Boiling Point 286°C to 287°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula C6H5CH2OC6H5
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 184.24
Percent Purity 98%
Physical Form Chunks
Chemical Name or Material Benzyl phenyl ether
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl phenyl ether
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'