missing translation for 'onlineSavingsMsg'
Learn More

gamma-Benzyl L-glutamate, 99%, ACROS Organics™

£17.25 - £159.24

Chemical Identifiers

CAS 1676-73-9
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.25
MDL Number MFCD00002633
InChI Key BGGHCRNCRWQABU-JTQLQIEISA-N
Synonym h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
PubChem CID 122337
IUPAC Name (2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10424345
SDS View Documents
Acros Organics
225670010
1g Glass bottle
£17.25
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 4
Please to purchase this item. Need a web account? Register with us today!
10022820
SDS View Documents
Acros Organics
225670050
5g Glass bottle
£51.01
5g
View alternative products
Save Up to 
»
Estimated Shipment: 10-08-2021
Please to purchase this item. Need a web account? Register with us today!
10157503
SDS View Documents
Acros Organics
225670250
25g Glass bottle
£159.24
25g
View alternative products
Save Up to 
»
In stock, ready to ship: 1
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

1676-73-9
237.25
BGGHCRNCRWQABU-JTQLQIEISA-N
122337
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N
C12H15NO4
MFCD00002633
h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
(2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid

Specifications

γ-Benzyl L-glutamate
Glass bottle
+ 27.2
162°C to 167°C
99+%
98.5% min. (HPLC)
C6H5CH2O2CCH2CH2CH(NH2)CO2H
h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N
237.25
237.25
99%
Authentic
+27.20° (20°C c=2,1N HCL)
White
1g
1676-73-9
C12H15NO4
MFCD00002633
BGGHCRNCRWQABU-JTQLQIEISA-N
(2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
122337
Powder
SDS
Product Certifications