missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloride, 99%, pure, stabilized, Thermo Scientific Chemicals

Catalog Number 10592052
13.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy
100-44-7, 75-56-9
MFCD00000889
KCXMKQUNVWSEMD-UHFFFAOYSA-N
benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluol
7503
CHEBI:615597
chloromethylbenzene
C1=CC=C(C=C1)CCl
This item is not returnable. View return policy
100-44-7, 75-56-9
MFCD00000889
KCXMKQUNVWSEMD-UHFFFAOYSA-N
benzyl chloride, chloromethyl benzene, benzylchloride, alpha-chlorotoluene, benzylchlorid, chlorophenylmethane, benzene, chloromethyl, chlorure de benzyle, tolyl chloride, alpha-chlortoluol
7503
CHEBI:615597
chloromethylbenzene
C1=CC=C(C=C1)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C7H7Cl
Linear Formula C6H5CH2Cl
Refractive Index 1.5370 to 1.5390 (20°C, 589nm)
Quantity 5 g
Stabilizer 0.25% propylene oxide
Molecular Weight (g/mol) 126.583
Formula Weight 126.59
Percent Purity ≥98.5%
Grade Pure
Physical Form Liquid
Chemical Name or Material Benzyl chloride, Stabilized, Pure
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzyl chloride, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'