missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol NF, Macron Fine Chemicals™

macron

Brand:  Macron Fine Chemicals 1403-12

Additional Details : Weight : 1.00000kg

Packaging Glass Bottle
Quantity 1L

 View more versions of this product

Product Code. 15587904

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

100-51-6
108.14
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
CHEBI:17987
C1=CC=C(C=C1)CO
C7H8O
WVDDGKGOMKODPV-UHFFFAOYSA-N
244
phenylmethanol

Specifications

Benzyl Alcohol
1L
C7H8O
WVDDGKGOMKODPV-UHFFFAOYSA-N
phenylmethanol
244
108.14
Glass Bottle
100-51-6
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
C1=CC=C(C=C1)CO
108.14
CHEBI:17987
NF
Videos
SDS
Documents