missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Alcohol NF, Macron Fine Chemicals™
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass
Glass Bottle
Specifications
Packaging | Glass Bottle |
Quantity | 1 L |
Formula Weight | 108.14 |
Grade | NF |
Chemical Name or Material | Benzyl Alcohol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'