missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, Multi-Compendial, N.F., J.T. Baker™

Catalog Number 15508514
154.85 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
100 mL
1 L
4 L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
This item is not returnable. View return policy
100-51-6
C7H8O
108.14
MFCD00004599,MFCD03792087
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
244
CHEBI:17987
phenylmethanol
OCC1=CC=CC=C1
This item is not returnable. View return policy
100-51-6
C7H8O
108.14
MFCD00004599,MFCD03792087
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
244
CHEBI:17987
phenylmethanol
OCC1=CC=CC=C1
Flash Point 93.00°C
Packaging Glass Bottle
Quantity 100 mL
Specific Gravity 1L = 1.05kg
Formula Weight 108.14
Grade Multi-Compendial/NF
Chemical Name or Material Benzyl Alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'