missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5391069
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
100-51-6
C7H8O
108.14
MFCD00004599,MFCD03792087
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
244
CHEBI:17987
phenylmethanol
OCC1=CC=CC=C1
This item is not returnable. View return policy
100-51-6
C7H8O
108.14
MFCD00004599,MFCD03792087
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
244
CHEBI:17987
phenylmethanol
OCC1=CC=CC=C1

High quality chemicals for laboratory and specialized industrial use

Flash Point 93.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1L = 1.05kg
Formula Weight 108.14
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Benzyl Alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'