missing translation for 'onlineSavingsMsg'
Learn More

Benzyl alcohol, for analysis, Thermo Scientific Chemicals

Catalog Number p-3523369
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
100-51-6
C7H8O
108.14
MFCD00004599,MFCD03792087
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
244
CHEBI:17987
phenylmethanol
OCC1=CC=CC=C1
100-51-6
C7H8O
108.14
MFCD00004599,MFCD03792087
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
244
CHEBI:17987
phenylmethanol
OCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY
Alkalinity 0.002 meq/g max.
Acidity 0.001 meq/g max.
Melting Point -15.0°C
Density 1.0440g/mL
Boiling Point 205.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH2OH
Refractive Index 1.5380 to 1.5410
Quantity 1 L
Merck Index 15, 1127
Specific Gravity 1.044
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: miscible with alcohol,ether,acetone,chloroform,,dmf and methylene chloride
Viscosity 6.6 mPa.s (20°C)
Formula Weight 108.14
Percent Purity ≥99.5%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Benzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'